Would you like to log in again?
QuantiChrom™ Albumin (BCG) Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
DIAG-250
75877-980EA 501.48 USD InStock
75877-980
QuantiChrom™ Albumin (BCG) Assay Kit, BioAssay Systems
Assays Protein Assays
For quantitative determination of albumin concentrations (BCG method).
  • Use as little as 5 μL samples. Detection range 0.01 - 5 g/dL (1.5 - 750 μM) albumin in 96-well plate assay
  • Can be readily automated to assay thousands of samples per day
Albumin is the most abundant plasma protein in human
BioAssay Systems’ BCG albumin assay kit is designed to measure albumin directly in biological samples without any pretreatment. The improved method utilizes bromcresol green that forms a colored complex specifically with albumin. The intensity of the color, measured at 620nm, is directly proportional to the albumin concentration in the sample. The optimized formulation substantially reduces interference by substances in the raw samples.
It accounts for about 60% of the total serum protein. Albumin plays important physiological roles, including maintenance of colloid osmotic pressure, binding of key substances such as long-chain fatty acids, bile acids, bilirubin, haematin, calcium and magnesium. It has anti-oxidant and anticoagulant effects, and also acts as a carrier for nutritional factors and drugs, as an effective plasma pH buffer. Serum albumin is a reliable prognostic indicator for morbidity and mortality, liver disease, nephritic syndrome, malnutrition and protein-losing enteropathies. High levels are associated with dehydration. Simple, direct and automation-ready. Procedures for measuring albumin concentration in biological samples are becoming popular in Research and Drug Discovery.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty