Would you like to log in again?
QuantiChrom™ Heme Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
DIHM-250
75877-998EA 539.69 USD InStock
75877-998
QuantiChrom™ Heme Assay Kit, BioAssay Systems
Assays Immunological Assays
For quantitative determination of heme and evaluation of drug effects on heme metabolism.
  • Safe
  • Reagents are non-toxic
  • Linear detection range 0.6 - 125 μM heme
  • The "mix-and-read" 5 minute procedure
  • Can be readily automated to process tens of thousands of samples per day
  • Assays can be executed in microplate or cuvet
Heme is one important member of the porphyrin family. It is synthesized in both mitochondria and cytoplasm, and is a key prosthetic group for various essential proteins such as hemoglobin, cytochromes, catalases and peroxidases. Heme determination is widely practiced by researchers of various blood diseases. Simple, direct and automation-ready. Procedures for measuring heme concentration are becoming popular in Research and Drug Discovery. BioAssay Systems’ QuantiChrom™ Heme Assay Kit is based on an improved aqueous alkaline solution method, in which the heme is converted into a uniform colored form. The intensity of color, measured at 400 nm, is directly proportional to the heme concentration in the sample. The optimized formulation substantially reduces interference by substances in the raw samples and exhibits high sensitivity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty