Would you like to log in again?
QuantiChrom™ Magnesium Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
DIMG-250
75878-000EA 588.41 USD InStock
75878-000
QuantiChrom™ Magnesium Assay Kit, BioAssay Systems
Assays Enzyme Assays
For quantitative determination of Mg2+ and evaluation of drug effects on magnesium metabolism.
  • Linear detection range 0.1 - 3 mg/dL (41 - 1200 uM) Mg2+ in 96-well plate assay
  • Can readily automated to process thousands of samples per day
  • Assays can be executed in microplate or cuvet
Magnesium (Mg) is one of the most abundant and essential minerals in mammals
BioAssay Systems’ magnesium assay kit is designed to measure magnesium directly in biological samples without any pretreatment. A calmagite dye in the kit forms a colored complex specifically with magnesium. The intensity of the color, measured at 500 nm, is directly proportional to the magnesium concentration in the sample. The optimized formulation minimizes interference by potential substances.
Magnesium is involved in more than 300 biochemical reactions in the body and plays important roles in muscle and nerve functions, heart rhythm, immune system and bone formation. Magnesium deficiency may lead to nausea, fatigue, muscle contractions, hypocalcemia and hypokalemia. Simple, direct and automation-ready. Procedures for measuring magnesium concentration in biological samples are becoming popular in Research and Drug Discovery.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty