Would you like to log in again?
QuantiChrom™ Phosphate Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
DIPI-500
75878-004EA 502.03 USD InStock
75878-004
QuantiChrom™ Phosphate Assay Kit, BioAssay Systems
Assays Cellular Assays
For quantitative determination of phosphate concentrations.
  • Linear detection range 0.3 - 50 μM (0.0028 - 0.47 mg/dL) phosphate in 96-well plate assay
  • Can readily automated for processing thousands of samples per day
Phosphate (Pi) is one of the most important ion species in nature
BioAssay Systems’ phosphate assay kit is designed to measure phosphate ion directly in samples without any pretreatment. The improved Malachite Green method utilizes the malachite green dye and molybdate, which forms a stable colored complex specifically with inorganic phosphate. The intensity of the color, measured at 620nm, is directly proportional to the phosphate concentration in the sample. The optimized formulation substantially reduces interference by substances in the raw samples.
Phosphate is present in all biological systems. It is a major constituent in minerals and fertilizers, and is a component of industrial wastewater. Thus accurate determination of phosphate concentration finds numerous applications in pharmacology, biomedical research, clinical chemistry, industrial process monitoring and environmental monitoring. Simple, direct and automation-ready. Procedures for measuring phosphate concentration in biological and environmental samples are becoming popular.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty