Would you like to log in again?
QuantiFluo™ Ammonia/Ammonium Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
DNH3-200
75878-008EA 245.86 USD InStock
75878-008
QuantiFluo™ Ammonia/Ammonium Assay Kit, BioAssay Systems
Assays Enzyme Assays
For quantitative fluorometric determination of ammonia/ammonium concentration.
  • Linear detection range of 0.012 - 1 mM ammonia
  • Can be readily automated to process thousands of samples per day
Ammonia (NH3) or its ion form ammonium (NH4+) is an important source of nitrogen for living systems and is ubiquitously present in the nature
BioAssay Systems’ ammonia/ammonium assay is based on an improved o-phthalaldehyde method. This reagent reacts with ammonia/ ammonium and forms a fluorescent product. The fluorescence intensity (λex/em = 360/450nm) is proportional to the ammonia concentration in the sample.
Simple, direct and automation-ready. Procedures for measuring NH3 are very desirable.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty