Would you like to log in again?
QuantiChrom™ Urease Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
DURE-100
75878-014EA 605.47 USD InStock
75878-014
QuantiChrom™ Urease Assay Kit, BioAssay Systems
Assays Enzyme Assays
For quantitative determination of urease enzyme activity.
  • Safe
  • Non-radioactive assay
  • As low as 0.003 U/L urease activity can be quantified
  • This "mix-incubate-measure" type assay can be readily automated to process thousands of samples per day
Urease (Amidohydrolase, EC 3.5.1.5) is an enzyme that catalyzes the hydrolysis of urea into carbon dioxide and ammonia
BioAssay Systems’ Urease Assay Kit provides a very sensitive and convenient means to measure urease activity in a variety of samples including soil. In the assay, urease reacts with urea, resulting in the formation of ammonia, which is determined by the Berthelot method at 670nm. The assay is simple, sensitive, stable and high-throughput adaptable.
(NH2)2)CO + H2O → CO2 + 2NH3) Many gastrointestinal or urinary tract pathogens produce urease. Thus its activity is a useful diagnostic parameter for the presence of pathogens such as Helicobacter pylori. Urease is found in bacteria, yeast, and higher plants. Urease activity is commonly determined in anaerobes of the bovine rumen, human feces and environmental samples such as soils and phytoplanktons.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty