Would you like to log in again?
EnzyChrom™ ADP Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
E2ADP-100
75878-018EA 585.08 USD InStock
75878-018
EnzyChrom™ ADP Assay Kit, BioAssay Systems
Assays Cellular Assays
For quantitative fluorometric determination of ADP determination in cells and other biological samples.
  • Safe and sensitive
  • Non-radioactive assay
  • As low as 0.1 μM ADP can be quantified
  • Can be readily automated to process thousands of samples per day
Adenosine diphosphate (ADP) is the product of ATP dephosphorylation by ATPases
BioAssay Systems’ newly designed ADP Assay Kit provides a convenient fluorometric means to measure ADP level even in the presence of ATP. In the assay, ADP is converted to ATP and pyruvate. The generated pyruvate is then quantified by a fluorimetric method (λex/em = 530/590nm). The assay is simple, sensitive, stable, high-throughput adaptable and can detect as low as 0.1 μM ADP in biological samples.
ADP can be converted back to ATP by ATP synthases. ADP levels regulate several enzymes involved in intermediary metabolism. Conventionally, ADP levels are measured by luciferase/luciferin mediated assays after ADP is converted to ATP. However, since these assays require measurement of ATP in the sample before conversion of ADP to ATP, if the nascent ATP concentration is significantly higher than the ADP concentration, the ATP signal will drown out the ADP signal.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty