Would you like to log in again?
EnzyChrom™ Glycogen Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
E2GN-100
75878-022EA 611.52 USD InStock
75878-022
EnzyChrom™ Glycogen Assay Kit, BioAssay Systems
Assays Cellular Assays
For quantitative determination of glycogen and evaluation of drug effects on glycogen metabolism.
  • Linear detection range 2 to 200 μg/mL glycogen for colorimetric assays and 0.2 to 20 μg/mL for fluorimetric assays
Glycogen is a branched polysaccharide of glucose units linked by α-1,4 glycosidic bonds and α-1,6 glycosidic bonds
BioAssay Systems’ glycogen assay uses a single Working Reagent that combines the enzymatic break down of glycogen and the detection of glucose in one step. The color intensity of the reaction product at 570nm or fluorescence intensity at λex/em = 530/585 nm is directly proportional to the glycogen concentration in the sample. This simple convenient assay is carried out at room temperature and takes only 30 min.
It is stored primarily in the liver and muscle, and forms an energy reserve that can be quickly mobilized to meet a sudden need for glucose. The most common glycogen metabolism disorder is found in diabetes, in which, due to abnormal amounts of insulin, liver glycogen can be abnormally accumulated or depleted. Genetic glycogen storage diseases have been associated with various inborn errors of metabolism caused by deficiencies of enzymes necessary for glycogen synthesis or breakdown. Simple, direct and automation-ready. Procedures for measuring glycogen concentrations find wide applications in research and drug discovery.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty