Would you like to log in again?
EnzyLight™ ADP Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
EADP-100
75878-034EA 505.36 USD InStock
75878-034
EnzyLight™ ADP Assay Kit, BioAssay Systems
Assays Cellular Assays
For rapid, quantitative, bioluminescent determination of ADP concentration and evaluation of drug effects on ADP metabolism.
  • Safe
  • Non-radioactive assay
  • As low as 0.02 μM ADP can be quantified
  • Can be readily automated to process thousands of samples per day
BioAssay Systems’ EnzyLight™ ADP Assay Kit provides a rapid method to measure ADP levels. The assay involves two steps. In the first step, the working reagent lyses cells to release ATP and ADP. In the presence of luciferase, ATP immediately reacts with the Substrate D-luciferin to produce light. The light intensity is a direct measure of intracellular ATP concentration. In the second step, the ADP is converted to ATP through an enzyme reaction. This newly formed ATP then reacts with the D-luciferin as in the first step. The second light intensity measured represents the total ADP and ATP concentration in the sample. This non-radioactive, homogeneous cell-based assay is performed in microplates. The reagent is compatible with all culture media and with all liquid handling systems for high-throughput screening applications in 96-well and 384-well plates.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty