Would you like to log in again?
EnzyChrom™ Alanine Transaminase Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
EALT-100
75878-038EA 508.84 USD InStock
75878-038
EnzyChrom™ Alanine Transaminase Assay Kit, BioAssay Systems
Assays Enzyme Assays
For quantitative determination of alanine transaminase activity.
  • Linear detection range 2-100 U/L
  • Assay can be carried out in a microplate or a cuvette and takes only 10 min
Alanine transaminase (ALT), also known as serum alanine aminotransferase (ALAT) or pyruvic transaminase (SGPT), facilitates the conversion of alanine and a-ketoglutarate to pyruvate and glutamate
BioAssay Systems’ ALT activity assay is based on the quantification of pyruvate produced by ALT. In this assay, pyruvate and NADH are converted to lactate and NAD by the enzyme lactate dehydrogenase (LDH). The decrease in NADH absorbance at 340 nm is proportional to ALT activity.
ALT plays an important role in gluconeogenesis and amino acid metabolism. ALT is found mainly in the liver, and, to a lesser extent, in kidney, heart, muscle, and pancreas tissues. Normal serum levels of ALT are low, and increased serum ALT activity is widely used as a marker for liver damage. Simple, direct and automation-ready. Procedures for measuring ALT activity find wide applications in research and drug discovery.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty