Would you like to log in again?
EnzyChrom™ Aspartate Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
EASP-100
75878-046EA 571.96 USD InStock
75878-046
EnzyChrom™ Aspartate Assay Kit, BioAssay Systems
Assays Immunological Assays
For quantitative determination of aspartate and evaluation of drug effects on aspartate metabolism.
  • Linear detection range 2 to 400 μM aspartate for colorimetric assays and 1 to 50 μM for fluorimetric assays
Aspartate, a nonessential amino acid, is a precursor to several other amino acids and is an excitatory neurotransmitter
BioAssay Systems’ Aspartate Assay Kit provides a simple, direct and automation-ready procedure for measuring aspartate concentration. Aspartate is converted into pyruvate which is then oxidized with the conversion of the dye into a colored and fluorescent form. The color intensity of the oxidized dye at 570 nm or fluorescence intensity at λex/em = 530/585 nm is directly proportional to the aspartate concentration in the sample.
Aspartate is involved in the urea cycle and gluconeogenesis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty