Would you like to log in again?
EnzyLight™ ATP Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
EATP-100
75878-050EA 489.96 USD InStock
75878-050
EnzyLight™ ATP Assay Kit, BioAssay Systems
Assays Cellular Assays
For rapid, quantitative, bioluminescent determination of ATP and evaluation of drug effects on ATP metabolism.
  • Safe. Non-radioactive assay. As low as 0.02 μM ATP or a single cell can be quantified
  • Convenient "mix-incubate-measure" type assay. No wash and reagent transfer steps are involved
  • Can be readily automated on HTS liquid handling systems
Adenosine 5’-triphosphate (ATP) is the chemical energy for cellular metabolism and is often referred to as “energy currency" of the cell
BioAssay Systems’ EnzyLight™ ATP Assay Kit provides a rapid method to measure intracellular ATP. The single working reagent lyses cells to release ATP, which, in the presence of luciferase, immediately reacts with the Substrate D-luciferin to produce light. The light intensity is a direct measure of intracellular ATP concentration. This non-radioactive, homogeneous cell-based assay is performed in microplates. The reagent is compatible with all liquid handling systems for high-throughput screening applications in 96-well and 384-well plates.
ATP is produced only in living cells during photosynthesis and cellular respiration and consumed in cellular processes including biosynthetic reactions, motility and cell division. It is a key indicator of cellular activity and has been utilized as a measure of cell viability and cytotoxicity in research and drug discovery.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty