Would you like to log in again?
EnzyChrom™ Glucose Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
EBGL-100
75878-052EA 542.72 USD InStock
75878-052
EnzyChrom™ Glucose Assay Kit, BioAssay Systems
Assays Cellular Assays
For quantitative colorimetric/fluorometric determination of glucose and evaluation of drug effects on glucose metabolism.
  • Linear detection range 5 to 300 μM (90 μg/dL to 5.4 mg/dL) glucose for colorimetric assays and 1 to 30 μM for fluorimetric assays
Glucose (C6H12O6) is a key diagnostic parameter for many metabolic disorders
BioAssay Systems’ glucose assay kit uses a single Working Reagent that combines the glucose oxidase reaction and color reaction in one step. The color intensity of the reaction product at 570 nm or fluorescence intensity at λex/em = 530/585 nm is directly proportional to glucose concentration in the sample.
Increased glucose levels have been associated with diabetes mellitus, hyperactivity of thyroid, pituitary and adrenal glands. Decreased levels are found in insulin secreting tumors, myxedema, hypopituitarism and hypoadrenalism. Simple, direct and high-throughput assays for measuring glucose concentrations find wide applications in research and drug discovery.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty