Would you like to log in again?
EnzyChrom™ Catalase Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
ECAT-100
75878-056EA 479.12 USD InStock
75878-056
EnzyChrom™ Catalase Assay Kit, BioAssay Systems
Assays Enzyme Assays
For quantitative determination of catalase activity and evaluation of drug effects on catalase activity.
  • Linear detection range 0.2 to 5 U/L catalase activity
Catalase (EC 1.11.1.6), is an ubiquitous antioxidant enzyme that catalyzes the decomposition of hydrogen peroxide (H2O2) to water and oxygen
BioAssay Systems’ improved assay directly measures catalase degradation of H2O2 using a redox dye. The change in color intensity at 570nm or fluorescence intensity (λex/em = 530/585nm) is directly proportional to the catalase activity in the sample.
By preventing excessive H2O2 build up, catalase allows important cellular processes which produce H2O2 as a byproduct to occur safely. Deficiency in catalase activity has been associated with grey hair and peroxisomal disorder acatalasia. Simple, direct and high-throughput assays for catalase activity find wide applications.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty