Would you like to log in again?
EnzyChrom™ Choline Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
ECHO-100
75878-064EA 600.41 USD InStock
75878-064
EnzyChrom™ Choline Assay Kit, BioAssay Systems
Assays Immunological Assays
For quantitative determination of choline and evaluation of drug effects on choline metabolism.
  • Linear detection range colorimetric assay 1 to 100 μM, fluorimetric assay 0.2 to 10 μM choline
Choline and its metabolites play important roles in membrane structure integrity, cellular signaling and cholinergic neurotransmission
BioAssay Systems’ method provides a simple, direct and high-throughput assay for measuring choline in biological samples. In this assay, free choline is oxidized by choline oxidase to βine and H2O2 which reacts with a specific dye to form a pink colored product. The color intensity at 570nm or fluorescence intensity (530/585 nm) is directly proportional to the choline concentration in the sample.
Aberrant regulation in choline metabolism has been associated with mental illness such as anxiety.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty