Would you like to log in again?
EnzyChrom™ Isocitrate Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ECIC-100
75878-066EA 571.82 USD InStock
75878-066
EnzyChrom™ Isocitrate Assay Kit, BioAssay Systems
Assays Immunological Assays
For quantitative colorimetric determination of isocitrate and evaluation of drug effects on its metabolism.
  • Linear detection range 20 to 5000 μM for 10 min reaction
  • Homogeneous "mix-incubate-measure" type assay Can be readily automated to process thousands of samples per day
Isocitrate (Isocitric acid) is a substrate in the citric acid (TCA) cycle
BioAssay Systems’ isocitrate assay measures the NADPH generated from the oxidation of isocitrate. The NADPH converts the dye to an intense violet color with an absorption maximum at 565 nm. The increase in absorbance at 565 nm is directly proportional to the isocitrate concentration.
Isocitrate is formed by the isomerization of citrate catalyzed by the enzyme aconitase. Isocitrate is oxidized by isocitrate dehydrogenase producing α-ketoglutarate and generating NADPH. Isocitrate is commonly found in many fruits and vegetables and their processed products. Industrially, isocitrate is used as a marker to identify the quality and purity of fruit juices.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty