Would you like to log in again?
EnzyChrom™ Creatine Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
ECRT-100
75878-078EA 612.51 USD InStock
75878-078
EnzyChrom™ Creatine Assay Kit, BioAssay Systems
Assays Immunological Assays
For quantitative determination of creatine and evaluation of drug effects on creatine metabolism.
  • Linear detection range 4 to 1000 μM (colorimetric assay) or 0.5 to 50 μM (fluorimetric assay)
  • Simple "mix-and-measure" procedure allows reliable quantitation of creatine within 30 minutes
Creatinine is present in vertebrates and helps to supply energy to muscle
BioAssay Systems’ creatine assay is based on enzymatic reactions leading to formation of a pink colored product. The optical density at 570 nm or fluorescence intensity at λ ex/em = 530/585 nm is directly proportional to the creatine concentration in the sample.
In humans and animals, approximately half of creatine originates from food (mainly from fresh meat). Creatine supplementation has been investigated as a possible therapeutic approach for the treatment of muscular, neuromuscular, neurological and neurodegenerative diseases. Simple, direct and automation-ready. Procedures for measuring creatine are popular in research and drug discovery.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty