Would you like to log in again?
EnzyFluo™ Isocitrate Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
EFIC-100
75878-086EA 560.02 USD InStock
75878-086
EnzyFluo™ Isocitrate Assay Kit, BioAssay Systems
Assays Cellular Assays
For quantitative fluorimetric determination of isocitrate and evaluation of drug effects on its metabolism.
  • Linear detection range 0.6 to 500 μM for 10 min reaction
  • Homogeneous "mix-incubate-measure" type high-throughput assay
Isocitrate (Isocitric acid) is a substrate in the citric acid (TCA) cycle
BioAssay Systems’ isocitrate assay measures the NADPH generated from the oxidation of isocitrate. The NADPH reduces a probe into a highly fluorescent product. The fluorescence intensity of this product, measured at λex/em = 530/585 nm, is proportional to the isocitrate concentration in the sample.
Isocitrate is formed by the isomerization of citrate catalyzed by the enzyme aconitase. Isocitrate is oxidized by isocitrate dehydrogenase producing α-ketoglutarate and generating NADPH. Isocitrate is commonly found in many fruits and vegetables and their processed products. Industrially, isocitrate is used as a marker to identify the quality and purity of fruit juices.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty