Would you like to log in again?
EnzyFluo™ Lactate Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
EFLLC-100
75878-088EA 620.31 USD InStock
75878-088
EnzyFluo™ Lactate Assay Kit, BioAssay Systems
Assays Cellular Assays
For quantitative fluorimetric determination of L-lactate (L-lactic acid) and evaluation of drug effects on its metabolism.
  • Linear detection range 1 - 50 μM L-lactate in 96-well plate assay
  • Can be readily automated to test thousands of samples per day
Lactate is generated by lactate dehydrogenase (LDH) under hypoxic or anaerobic conditions
BioAssay Systems’ EnzyFluo™ lactate assay kit is based on lactate dehydrogenase catalyzed oxidation of lactate, in which the formed NADH reduces a probe into a highly fluorescent product. The fluorescence intensity of this product, measured at λex/em = 530/585 nm, is proportional to the lactate concentration in the sample.
Monitoring lactate levels is, therefore, a good indicator of the balance between tissue oxygen demand and utilization and is useful when studying cellular and animal physiology. Simple, direct and automation-ready. Procedures for measuring lactate concentration are very desirable.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty