Would you like to log in again?
EnzyChrom™ Fumarase Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
EFMR-100
75878-090EA 746.04 USD InStock
75878-090
EnzyChrom™ Fumarase Assay Kit, BioAssay Systems
Assays Enzyme Assays
For quantitative determination of fumarase enzyme activity.
  • Linear detection range 0.4 to 70 U/L
  • Homogeneous "mix-incubate-measure" type high-throughput assay
Fumarase (or fumarate hydratase) (EC 4.2.1.2) is an enzyme that catalyzes the reversible hydration/dehydration reaction of fumarate to malate
BioAssay Systems’ non-radioactive, colorimetric fumarase assay is based on the reduction of the tetrazolium salt MTT in a NADH-coupled enzymatic reaction to a reduced form of MTT which exhibits an absorption maximum at 565 nm. The increase in absorbance at 565 nm is proportional to the enzyme activity.
Fumarase exists in two isoforms: a cytosolic and mitochondrial form. In the citric acid cycle, it facilitates a transition step in the production of energy in the form of NADH. Fumarase deficiency in humans results in early brain development problems and is characterized by poor feeding, hypotonia, failure to thrive, etc.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty