Would you like to log in again?
EnzyFluo™ ERK Phosphorylation Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
EERK-100
75878-108EA 789.7 USD InStock
75878-108
EnzyFluo™ ERK Phosphorylation Assay Kit, BioAssay Systems
Assays Cellular Assays
For quantitative fluorescent immunoenzymatic assay of ERK1/2 phosphorylation status in cultured cells.
  • Safe
  • Non-radioactive assay
  • Total and pERK can be measured in the same sample
The mitogen-activated protein kinase (MAPK/ERK) pathway plays a key role in cell proliferation, differentiation and migration
BioAssay Systems’ cell-based ELISA measures dually phosphorylated ERK1/2 in whole cells and normalizes the signal to the total protein content. This simple and efficient assay eliminates the need for cell lysate preparation and can be used to study kinase signaling and the effects of kinase inhibitors on cells. In this assay, cells are grown in 96-well plates and treated with ligands or drugs. Cells are then fixed and permeabilized in the wells. ERK1/2 phosphorylation (pERK) using a fluorescent ELISA followed by total protein measurement in each well.
Stimulation by mitogens eventually leads to phosphorylation of ERK1 (T202/Y204) and ERK2 (T185/Y187). The MAPK/ERK cascade presents many interesting drug targets for the development of cancer therapies.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty