Would you like to log in again?
EnzyChrom™ Glucose-6-Phosphate Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
EG6P-100
75878-112EA 596.09 USD InStock
75878-112
EnzyChrom™ Glucose-6-Phosphate Assay Kit, BioAssay Systems
Assays Cellular Assays
For quantitative determination of glucose-6-phosphate in biological samples.
  • Linear detection range 10 to 1000 μM G6P
  • Homogeneous "mix-incubate-measure" type high-throughput assay
Glucose-6-Phosphate (g6p) is glucose sugar phosphorylated on carbon 6
BioAssay Systems’ Glucose-6-Phosphate Assay Kit provides a simple, and automation-ready procedure for measuring G6P concentration. G6P is oxidized by glucose-6-phosphate dehydrogenase and the formed NADPH is coupled to the formazan (WST-8) chromogen. The intensity of the product color, measured at 460 nm, is proportional to the G6P concentration in the sample.
Most of the glucose entering cells is phosphorylated to G6P. G6P has three primary fates within the cell. It lies at the start of two major metabolic pathways: glycolysis and the pentose phosphate pathway. In addition to these metabolic pathways, glucose 6-phosphate may also be converted to glycogen or starch for storage.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty