Would you like to log in again?
EnzyLight™ ADP/ATP ratio Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
ELDT-100
75878-114EA 547.6 USD InStock
75878-114
EnzyLight™ ADP/ATP ratio Assay Kit, BioAssay Systems
Assays Cellular Assays
For quantitative bioluminescent assay for ADP:ATP ratio (apoptosis) in cells.
  • Safe
  • Non-radioactive assay
  • Homogeneous and convenient "mix-incubate-measure" type assay
  • Can be readily automated to test thousands of samples per day
Changes in the ADP/ATP ratio have been used to differentiate modes of cell death and viability
BioAssay Systems’ EnzyLight™ ADP/ATP Ratio Assay Kit provides a rapid method to measure ADP and ATP levels for the screening of apoptosis, necrosis and cell proliferation in mammalian cells. The assay involves two steps. In the first step, the working reagent lyses cells to release ATP and ADP. In the presence of luciferase, ATP immediately reacts with the Substrate D-luciferin to produce light. The light intensity is a direct measure of intracellular ATP concentration. In the second step, the ADP is converted to ATP through an enzyme reaction. This newly formed ATP then reacts with the D-luciferin as in the first step. This non-radioactive, homogeneous cell-based assay is performed in microplates. The reagent is compatible with all culture media and with all liquid handling systems for high-throughput screening applications in 96-well and 384-well plates.
Increased levels of ATP and decreased levels of ADP signify proliferating cells. Conversely, decreased levels of ATP and increased levels of ADP represent apoptotic or necrotic cells where the decrease in ATP and increase in ADP is much more pronounced in necrosis versus apoptosis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty