Would you like to log in again?
QuantiFluo™ Alkaline Phosphatase Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
QFAP-100
75878-120EA 317.41 USD InStock
75878-120
QuantiFluo™ Alkaline Phosphatase Assay Kit, BioAssay Systems
Assays Enzyme Assays
For quantitative fluorometric determination of alkaline phosphatase (ALP) activity.
  • Detection limit of 0.02 U/L
  • Simple "mix-and-measure" procedure allows reliable quantitation of ALP activity within 20 minutes
  • All reagents are compatible with high-throughput liquid handling instruments
Alkaline phosphatase (ALP) catalyzes the hydrolysis of phosphate esters in an alkaline environment, resulting in the formation of an organic radical and inorganic phosphate. In mammals, this enzyme is found mainly in the liver and bones. Marked increase in serum ALP levels, a disease known as hyperalkalinephosphatasemia, has been associated with malignant biliary obstruction, primary biliary cirrhosis, primary sclerosing cholangitis, hepatic lymphoma and sarcoidosis. Simple, direct and automation-ready. Procedures for measuring ALP activity in serum are becoming popular in Research and Drug Discovery. This improved method utilizes 4-methylumbelliferyl phosphate that is hydrolyzed by ALP into a highly fluorescent product 4-methylumbelliferone. The rate of the fluorescence increase is directly proportional to the enzyme activity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty