Would you like to log in again?
QuantiFluo™ DNA Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
QFDN-250
75878-122EA 486.31 USD InStock
75878-122
QuantiFluo™ DNA Assay Kit, BioAssay Systems
Assays Cellular Assays
For quantitative determination of DNA.
  • Linear detection range 2 - 40 ng (100 - 2,000 ng/mL) calf thymus DNA in 96-well plate assay
  • Convenient
  • Single working reagent
  • Can readily automated to test tens of thousands of samples per day
  • Low interference. RNA, salt (up to 3M NaCl), detergent (< 0.01% SDS) and common DNA extraction buffer do not interfere in the assay
DNA quantitation is a common practice in molecular biology
BioAssay Systems’ QuantiFluo™ DNA assay kit is designed to accurately measure nanogram quantities of plasmid DNA, cDNA, DNA following polymerase chain reaction and DNA eluted from gels. The improved method utilizes Hoechst dye that binds specifically with double-stranded DNA. The fluorescence intensity, measured at λex/em = 350/450nm, is directly proportional to the DNA concentration in the sample. The optimized formulation substantially reduces interference by substances in the raw samples.
Very often DNA is available in minute quantities and the traditional UV 260 nm absorbance method requires microgram quantities for reliable results. Accurate determination of DNA concentration, especially when DNA is present at low concentrations, is crucial for reproducible results in sequencing, cloning, transfection and DNA labeling. Simple, direct and automation-ready. Procedures for measuring DNA concentration are very desirable.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty