Would you like to log in again?
EnzyChrom™ Glycerol Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
EGLY-200
75878-132EA 590.03 USD InStock
75878-132
EnzyChrom™ Glycerol Assay Kit, BioAssay Systems
Assays Immunological Assays
For quantitative determination of glycerol and evaluation of drug effects on glycerol metabolism.
  • Linear detection range 10 to 1000 μM (92 μg/dL to 9.2 mg/dL) glycerol for colorimetric assays and 2 to 50 μM for fluorimetric assays
  • Homogeneous high-throughput assay
Glycerol [Glycerin or Glycerine, C3H5 (OH) 3] is widely used in foods, beverages and pharmaceutical formulations
BioAssay Systems’ glycerol assay uses a single Working Reagent that combines glycerol kinase, glycerol phosphate oxidase and color reactions in one step. The color intensity of the reaction product at 570nm or fluorescence intensity at λex/em = 530/585nm is directly proportional to glycerol concentration in the sample.
It is also a main by-product of biodiesel production. Simple, direct and automation-ready. Procedures for measuring glycerol concentrations find wide applications.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty