Would you like to log in again?
EnzyChrom™ Glucose Oxidase Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
EGOX-100
75878-134EA 540.62 USD InStock
75878-134
EnzyChrom™ Glucose Oxidase Assay Kit, BioAssay Systems
Assays Enzyme Assays
For quantitative determination of glucose oxidase activity and evaluation of drug effects on its metabolism.
  • Linear detection range 0.02 to 10 U/L glucose oxidase for colorimetric assays and 0.002 to 1.5 U/L for fluorimetric assays
  • Homogeneous high-throughput assay
Glucose oxidase catalyzes the oxidation of glucose from D-glucose to D-glucono-δ-lactone
BioAssay Systems’ glucose oxidase assay kit uses a single Working Reagent that combines the glucose oxidase reaction and color reaction in one step. The change in color intensity of the reaction product at 570 nm or fluorescence intensity at λex/em = 530/585 nm is directly proportional to glucose oxidase activity in the sample.
Physiologically, it aids in the breakdown of glucose into smaller metabolites. It is widely used in electrochemical glucose sensors designed for diabetes patients. Simple, direct and high-throughput assays for measuring glucose oxidase activity find wide applications in research and drug discovery.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty