Would you like to log in again?
EnzyChrom™ Invertase/Sucrase Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
EIVT-100
75878-142EA 663.23 USD InStock
75878-142
EnzyChrom™ Invertase/Sucrase Assay Kit, BioAssay Systems
Assays Enzyme Assays
For quantitative determination of invertase/sucrase activity.
  • Safe
  • Non-radioactive assay
  • "Mix-incubate-measure" type assay. No wash and reagent transfer steps are involved
  • Can be readily automated to test thousands of samples per day
Invertase (b-fructofuranosidase, EC 3.2.1.26) is an enzyme that catalyzes the hydrolysis of sucrose to fructose and glucose
BioAssay Systems’ Invertase Assay Kit provides a convenient and ultra-sensitive colorimetric and fluorimetric means to measure invertase activity. In the assay, invertase cleaves sucrose, resulting in the formation of fructose and glucose, which is determined by a colorimetric (570nm) or fluorimetric method (λex/em = 530/585nm). The assay is simple, sensitive, stable and high-throughput adaptable.
Invertases cleave at the O-C(fructose) bond, whereas a related enzyme sucrase (EC 3.2.1.48) cleaves at the O-C(glucose) bond. A wide range of microorganisms produce invertase and can, thus, utilize sucrose as a nutrient. Invertase assay finds wide applications in environmental (e.g. soil), agricultural and food (confectionery) industry.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty