Would you like to log in again?
EnzyChrom™ Nitric Oxide Synthase Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
ENOS-100
75878-158EA 620.45 USD InStock
75878-158
EnzyChrom™ Nitric Oxide Synthase Assay Kit, BioAssay Systems
Assays Enzyme Assays
For quantitative determination of nitric oxide synthase (NOS) activity.
  • Detection range 0.25 - 25 U/L in 96-well plate
  • Rapid and reliable
Nitric oxide (NO) is a reactive radical that plays an important role in many key physiological functions
BioAssay Systems’ EnzyChrom™ Nitric Oxide Synthase Assay Kit involves two steps: a NOS reaction step during which NO is produced followed by an NO detection step. Since the NO generated by NOS is rapidly oxidized to nitrite and nitrate, the NO production is measured following reduction of nitrate to nitrite using an improved Griess method. The procedure is simple and the time required for sample pretreatment and assay is reduced to as short as 40 min.
NO, an oxidation product of arginine by nitric oxide synthase (NOS), is involved in host defense and development, activation of regulatory proteins and direct covalent interaction with functional biomolecules. Simple, direct and non-radioactive. Procedures for measuring NOS are becoming popular in Research and Drug Discovery.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty