Would you like to log in again?
EnzyChrom™ Phospholipase D Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
EPPD-100
75878-162EA 520.73 USD InStock
75878-162
EnzyChrom™ Phospholipase D Assay Kit, BioAssay Systems
Assays Enzyme Assays
For quantitative determination of phospholipase D activity.
  • Detection range colorimetric assay 0.06 - 10 U/L, fluorimetric assay 0.04 - 1 U/L
  • Can be readily adapted to high-throughput assays for drug screening
Phospholipase D (PLD) catalyses the hydrolysis of the phosphodiester bond of glycerophospholipids to generate phosphatidic acid and a free headgroup
BioAssay Systems’ method provides a simple and high-throughput assay for measuring PLD activity. In this assay, PLD hydrolyzes phosphatidylcholine to choline which is determined using choline oxidase and a H2O2 specific dye. The optical density of the pink colored product at 570nm or fluorescence intensity (530/585 nm) is directly proportional to the PLD activity in the sample.
Abnormalities in PLD expression have been associated with human cancers.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty