Would you like to log in again?
EnzyChrom™ Sialic acid Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
ESLA-100
75878-170EA 711.05 USD InStock
75878-170
EnzyChrom™ Sialic acid Assay Kit, BioAssay Systems
Assays Immunological Assays
For quantitative determination of free and total sialic acid (NANA) concentration.
  • Linear detection range 20 - 1000 μM sialic acid for colorimetric assays and 2 - 100 μM for fluorimetric assays
  • Can detect both free and total sialic acid
Sialic acid is a general name for nine carbon acidic sugars with N- or O-substituted derivatives
BioAssay Systems’ sialic acid assay uses a single Working Reagent that combines NANA aldolase, pyruvate oxidase and hydrogen peroxide determination in one step. The color intensity of the reaction product at 570nm or fluorescence intensity at λex/em = 530/585nm is directly proportional to sialic acid concentration in the sample.
The most common member of these sugars is N-acetylneuraminic acid (NANA). Sialic acid is widely distributed throughout mammalian tissues and fluids including serum. Sialylated oligosaccharides have been shown to exhibit antiviral properties and are also known to influence blood coagulation and cholesterol levels. The Sialic acid level in body fluids is also an important marker for diagnosing cancer. Simple, direct and automation-ready. Procedures for measuring sialic acid concentrations find wide applications in research and drug discovery.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty