Would you like to log in again?
EnzyChrom™ Superoxide Dismutase Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
ESOD-100
75878-172EA 488.31 USD InStock
75878-172
EnzyChrom™ Superoxide Dismutase Assay Kit, BioAssay Systems
Assays Enzyme Assays
For quantitative determination of superoxide dismutase (SOD) enzyme activity.
  • Linear detection range of 0.05 - 3 U/mL SOD
  • No wash and reagent transfer steps are involved
  • Homogeneous "mix-incubate-measure" type high-throughput assay
Superoxide dismutases (SOD, EC1.15.1.1) are enzymes that catalyze the dismutation of superoxide into O2 and H2O2
BioAssay Systems’ SOD assay provides a convenient colorimetric means for the quantitative determination of SOD enzyme activity in biological samples. In the assay, superoxide (O2-) is provided by xanthine oxidase (XO) catalyzed reaction. O2- reacts with a WST-1 dye to form a colored product. SOD scavenges the O2- thus less O2- is available for the chromogenic reaction. The color intensity (OD440nm) is used to determine the SOD activity in a sample.
They are an important antioxidant defense in all cells exposed to O2. There are three major families of superoxide dismutase: Cu/Zn, Fe/Mn, and the Ni type. Aberrant SOD activities have been linked to diseases such as amyotrophic lateral sclerosis, perinatal lethality, neural disorders and cancer.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty