Would you like to log in again?
EnzyChrom™ Triglyceride Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
ETGA-200
75878-176EA 612.65 USD InStock
75878-176
EnzyChrom™ Triglyceride Assay Kit, BioAssay Systems
Assays Immunological Assays
For quantitative determination of triglyceride and evaluation of drug effects on triglyceride metabolism.
  • Linear detection range 0.01 - 1.0 mmol/L (0.88 - 88.5 mg/dL) triglyceride
  • Convenient room temperature high-throughput assay
Triglyceride, also known as Triacyltriglyceride or Triacyl-glyceride, is the main constituent in vegetable oil and animal fats
BioAssay Systems’ triglyceride assay uses a single Working Reagent that combines triglyceride hydrolysis and glycerol determination in one step, in which a dye reagent is oxidized to form a colored product. The color intensity at 570nm is directly proportional to triglyceride concentration in the sample.
Triglycerides play an important role as energy sources and transporters of dietary fat. In the human body, high levels of triglycerides in the bloodstream have been linked to atherosclerosis, heart disease and pancreatitis. Simple, direct and automation-ready. Procedures for measuring triglyceride concentrations find wide applications in research and drug discovery.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty