Would you like to log in again?
EnzyChrom™ Xanthine Oxidase Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
EXOX-100
75878-180EA 491.83 USD InStock
75878-180
EnzyChrom™ Xanthine Oxidase Assay Kit, BioAssay Systems
Assays Enzyme Assays
For quantitative determination of xanthine oxidase enzyme activity.
  • Linear detection range 0.03 - 25 U/L xanthine oxidase for colorimetric assays and 0.01 -2.5 U/L for fluorimetric assays
  • Convenient room temperature and high-throughput assay
Xanthine oxidase catalyzes the oxidation of xanthine to uric acid
BioAssay Systems’ xanthine oxidase assay kit uses a single Working Reagent that combines the xanthine oxidase reaction and color reaction in one step. The change in color intensity of the reaction product at 570 nm or fluorescence intensity at λex/em = 530/585 nm is directly proportional to xanthine oxidase activity in the sample.
In addition, xanthine oxidase can catalyze the oxidation of hypoxanthine to xanthine, act on certain purines and aldehydes, and in certain cases produce the superoxide ion. Clinically, xanthine oxidase activity in blood can act as a marker for influenza, liver damage, and possibly cardiovascular health. Simple, direct and high-throughput assays for measuring xanthine oxidase activity find wide applications in research and drug discovery.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty