Would you like to log in again?
Malachite G Phosphate Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
POMG-25H
75878-186EA 381.14 USD InStock
75878-186
Malachite G Phosphate Assay Kit, BioAssay Systems
Assays Enzyme Assays
For sensitive and high-throughput phosphate determination.
  • Very stable reagent
  • High sensitivity and wide detection range (0.02 - 40 μM phosphate)
  • Homogeneous "mix-and-measure" type high-throughput assay
  • Assays can be performed in tubes, cuvettes or microplates
The Malachite Green Phosphate Assay Kit is based on quantification of the green complex formed between Malachite Green, molybdate and free orthophosphate. The rapid color formation from the reaction can be conveniently measured on a spectrophotometer (600 - 660 nm) or on a plate reader. The non-radioactive colorimetric assay kits have been optimized to offer superior sensitivity and prolonged. Shelf Life. The assay is simple and fast, involving a single addition step for phosphate determination. Assays can be executed in tubes, cuvettes or multi-well plates. The assays can be conveniently performed in 96- and 384-well plates for high-throughput screening of enzyme inhibitors.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty