Would you like to log in again?
QuantiFluo™ ALP Detection Reagent, BioAssay Systems
Supplier: BioAssay Systems
QFALP-6ML
75878-190EA 182.46 USD InStock
75878-190
QuantiFluo™ ALP Detection Reagent, BioAssay Systems
Protein Expression Protein Expression Systems
Stabilized 4-methylumbelliferyl phosphate (MUP) liquid substrate reagent for detecting alkaline phosphatase activity.
  • Stable single reagent
  • Highly sensitive fluorescence detection
  • Can be visualized under a UV light
QuantiFluo™ ALP Reagent is a stable 4-methylumbelliferyl phosphate (MUP) liquid substrate reagent formulated for detecting alkaline phosphatase ALP, e.g. in ELISA. This single reagent produces a highly fluorescent product (λex/em = 360/450 nm) when dephosphorylated by ALP. The fluorescent product can also be visualized under a UV light.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty