Would you like to log in again?
QuantiFluo™ Protein Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
QFPR-200
75878-192EA 226.07 USD InStock
75878-192
QuantiFluo™ Protein Assay Kit, BioAssay Systems
Assays Protein Assays
For quantitative fluorimetric determination of total protein.
  • Linear detection range of 0.05 - 200 μg/mL BSA.
  • Homogeneous "mix-incubate-measure" type assay Can be readily automated to test thousands of samples per day
The protein or peptide is known as the "building blocks of life" and is one of the most important macromolecules in life science
BioAssay Systems’ QuantiFluo™ protein assay kit is based on an improved o-phthalaldehyde method. This reagent reacts with primary amines in protein or peptide and forms a blue fluorescent product, allowing detection of nanograms of proteins. The fluorescence intensity (λex/em = 360/450nm) is proportional to the protein concentration in the sample.
Protein determination is a very common practice. Simple, direct and automation-ready. Procedures for measuring protein or peptide concentration are very desirable
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty