Would you like to log in again?
QuantiChrom™ Sorbitol Dehydrogenase Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
DSDH-100
75878-326EA 617.39 USD InStock
75878-326
QuantiChrom™ Sorbitol Dehydrogenase Assay Kit, BioAssay Systems
Assays Enzyme Assays
For quantitative determination of sorbitol dehydrogenase enzyme activity.
  • Linear detection range 0.1 - 125 U/L sorbitol dehydrogenase
  • Can be readily automated to process thousands of samples per day
Sorbitol dehydrogenase (SDH) is an enzyme that catalyzes the interconversion of sorbitol and fructose. Elevated blood serum SDH levels indicate liver damage; thus, SDH plays an important role in the diagnosis of liver disease, especially in combination with aminotransferases. SDH levels are also measured to evaluate diabetic complications such as proliferative diabetic retinopathy. BioAssay Systems’ non-radioactive, colorimetric SDH assay is based on the reduction of the tetrazolium salt MTT in a NADH-coupled enzymatic reaction to a reduced form of MTT which exhibits an absorption maximum at 565 nm. The increase in absorbance at 565 nm is directly proportional to the enzyme activity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty