Would you like to log in again?
Fumarate Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
EFUM-100
75878-342EA 584.6 USD InStock
75878-342
Fumarate Assay Kit, BioAssay Systems
Assays Immunological Assays
For quantitative determination of D-lactate (D-lactic acid) and evaluation of drug effects on its metabolism
  • Fast and sensitive. Use of 20 µL sample. Linear detection range 0.005 to 2 mM fumarate in 96-well plate assay.
  • The procedure involves adding a single working reagent, and reading the optical density after 30 minutes. Room temperature assay. No 37°C heater is needed.
  • Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day
  • Method: OD565nm
  • Samples: Food, beverage and other biological samples (e.g. cell lysate, tissue homogenate, serum)
FUMARATE , or Fumaric Acid, is one of the key components in TCA cycle and is used by cells to form ATP. Human skin when exposed to sunlight will naturally produce fumaric acid. Fumarate is used as an additive by the food and beverage industries. Fumaric acid esters are also used to treat psoriasis. Increased urinary fumarate may be due to impaired Krebs cycle function, a defect in the enzyme fumarase or mitochondrial function. BioAssay Systems’ fumarate assay kit is based on fumarase catalyzed hydration of fumarate to malate. The malate is then oxidized by malate dehydrogenase generating NADH which reduces a formazan (MTT) dye. The intensity of the product color, measured at 565 nm is proportional to the fumarate concentration in the sample.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty