Would you like to log in again?
ReadyRack™ Tryptic Soy Broth (TSB), Disposable Tube Rack
Supplier: Hardy Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ReadyRack™
K380BX K82BX K82BX
10436-342PK 496.81 USD InStock
CA10436-342 10436-342 75878-364
ReadyRack™ Tryptic Soy Broth (TSB), Disposable Tube Rack
Media for Microbiology Prepared Media for Microbiology
Tryptic Soy Broth (TSB) in a ReadyRack™ is for sterility testing and cultivation of microorganisms. TSB is also known as Soybean-Casein Digest Broth, USP.
  • For sterility testing in cleanrooms
  • Can be rinsed with alcohol
  • Avoid contamination
The ReadyRack is a plastic rack designed for cleanroom use and can be rinsed with alcohol.
Simply remove ReadyRack from box outside of cleanroom. Spray down the plastic bag with isopropyl alcohol, and allow to dry. Take into cleanroom and remove plastic bag. Spray down entire ReadyRack with isopropyl alcohol, and allow to dry.
It is a convenient way to bring media into a controlled area while avoiding outside contamination.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty