Would you like to log in again?
Tryptic Soy Broth (TSB), USP
Supplier: Hardy Diagnostics
U363
75878-368PK 325.15 USD InStock
75878-368
Tryptic Soy Broth (TSB), USP
Media for Microbiology Dehydrated Media for Microbiology
For sterility testing and the cultivation of microorganisms.
Tryptic Soy Broth is recommended for use as a general purpose medium for the isolation and cultivation of a wide variety of bacteria and fungi. Also known as Soybean-Casein Digest Broth, USP.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty