Would you like to log in again?
Vis-U-All High Temperature Sterilization Pouches, Steris
Supplier: Steris
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
884122 884713 884116 884510 884039 884118
75879-212CS 1596.36 USD InStock
75879-212 75879-222 75879-216 75879-214 75879-218 75879-220
Vis-U-All High Temperature Sterilization Pouches, Steris
Bags Sterilization Pouches
Color coded for fast identification, these purple Vis-U-All high temperature sterilization pouches are validated for use with ethylene oxide and steam sterilization.
  • Event related sterile storage for up to one year
  • Chevron styled opening enhances aseptic opening
  • Fold over self seal design promotes even sealing and ease
2.0 ml thick polypropylene-polyethylene film combined with 42 lbs. medical grade paper provide excellent tear and puncture resistance.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty