Would you like to log in again?
Forceps, Adson, Excelta Corp®
Supplier: Excelta
AF-2-SE AF-1-SE 10G-SA AF-4-SE AF-3-SE F-6-SE 10H-S-SE
75880-458EA 3.67 USD InStock
75880-458 75880-724 10066-444 75880-460 75880-462 89411-852 75880-464
Forceps, Adson, Excelta Corp®
Forceps
This Adson dissection forceps is used for holding and manipulating delicate tissue.
  • Wide serrated thumb area for ergonomics
  • Excellent for handling delicate tissue
  • Cleanroom safe, stainless steel
This Adson stainless steel tissue forceps is used for holding and manipulating delicate tissues. These forceps have a wide, flat thumb grasp area that is commonly serrated. The serrated tips provide more 'sure' grip. The satin finish reduces glare.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty