Would you like to log in again?
Tweezers, Diamond Coated Tips, Excelta Corp®
Supplier: Excelta
SS-SA-DC 00-SA-DC 5-SA-DC 7-SA-DC 3C-SA-DC 2A-SA-DC
75880-796EA 130.77 USD InStock
75880-796 75880-764 75880-522 75880-788 75880-778 75880-504
Tweezers, Diamond Coated Tips, Excelta Corp®
Forceps
Designed for handling SMD components - Coated tips add strength and longevity to the tips.
  • Biocompatible
  • Chemically inert to 350 °C
  • Prevents particle shedding
  • Cleanroom safe, stainless steel
  • Protects the tips from wear while providing elasticity
These unique 'Diamond Coated' high precision pointed tweezers are made of anti-magnetic Neverust® stainless steel. The diamond coated tips are biocompatible, chemically inert to 350°C, prevent particle shedding, cleanroom safe, and protect the tips from wear while providing elasticity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty