Would you like to log in again?
TWEEZERS STRIPPING CARBON STEEL 115MM
Supplier: EXCELTA CORPORATION
Stripping tweezer 115mm^ Stripping tweezer, Quality: Five star, Shape: 70 degree Angulated, Material: Carbon Steel, PTFE and other coated wire without nicking, With fame hardened edges, Static dissipative epoxy coating (10⁵ - 10⁶ 0hms/sq), Cutting Capacity: 30 AWG, Dimension: 10x2.5mm, Length: 115mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TWEEZERS STRIPPING CARBON STEEL 115MM 75880-536 15A-ST-PE-30 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty