Would you like to log in again?
Pliers, Stress Relief, Excelta
Supplier: Excelta
554TX-US 554TXC-US 954A 954B 907-88 907-88C 900-88B 554B-US 554A-US 909-4B 900-10 554AD-US 554EF-US 909-4 554F-US 900-247 554TB-US 554TXB-US
75880-652EA 638.61 USD InStock
75880-652 63043-046 63043-010 89411-956 75880-656 75880-668 89411-958 63043-040 75880-626 75880-660 63043-048 63042-588 63042-610 63042-612 63042-604 75880-628 76243-292 63042-596
Pliers, Stress Relief, Excelta
Pliers
Designed to form stress relief for transistor and resistor leads.
  • Stress relief on transistors, resistors
  • Ergonomic grips for comfort
  • Double leaf springs for ease of use
All of these forming pliers provide stress relief for transistor and resistor leads. They feature an ergonomically molded antimicrobial grip (10¹⁰ ohms/sq surface resistivity) with dual leaf springs. The tools are made of carbon steel.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty