Would you like to log in again?
PLIER CUT BEND LEAD CARBON STEEL 5.5IN
Supplier: EXCELTA CORPORATION
Plier Cut & Bend Lead^ Plier Cut & Bend Lead, Quality Level: 5stars, Material: Carbon Steel, Tip Size: Shear, Type of Cut: 0.032in max, Over All Length: 5.5in, Body Width: 0.43in, Body Height: 0.26in, Designed to cut component leads at 0.060in then bends the lead keeping the component in place
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PLIER CUT BEND LEAD CARBON STEEL 5.5IN 75880-666 930E-1' Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty