Would you like to log in again?
Cutters, Transverse End, Excelta
Supplier: Excelta
7173E 7174E 7273E 7274E
63042-812EA 115.56 USD InStock
63042-812 75880-702 75880-694 94024-370
Cutters, Transverse End, Excelta
Cutters
Thes cutters are ideal for horizontal cutting on a vertical plane in tight places.
  • Transverse cutting blades for difficult areas
  • 'Set-Screw' stop prevents over-travel
  • Induction hardened blades for longevity
  • Ergonomic handles and dual leaf springs
These Lazer Line® miniature tapered transverse optimum flush cutters with a cutting capacity of 0.001 to 0.020" soft wire is made of carbon tool steel with induction hardened cutting blades to RHC 63-65. The CNC machining allows the joint surfaces to be perfect to allow for long life. This 6" tool has a 40° angled tip with a set-screw to prevent over-travel while prolonging the life of the tool. The grips are soft cushioned foam and have a surface resistivity of 10¹⁰ ohms/sq. The dual leaf springs provide smooth action.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty