Would you like to log in again?
Forceps, Gemologist, Teflon Coated Tip, Excelta Corp®
Supplier: Excelta
JST-1 JST-2
75880-756EA 97.03 USD InStock
75880-756 75880-758
Forceps, Gemologist, Teflon Coated Tip, Excelta Corp®
Forceps
Designed for handling gem stones without marring.
  • Teflon coated tips avoid marring
  • Various sizes available
  • Stainless steel corrosion resistant
These gemologist straight medium point tweezers are ideal for handling medium gems. The handles are serrated for better control. They feature teflon coated tips to prevent scratching.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty