Would you like to log in again?
TWEEZERS STRONG MEDIUM PNT CARBOFB 4.5IN
Supplier: EXCELTA CORPORATION
Tweezer 4.5in^ Tweezer, The 00 three star straight medium pointed tweezer is the most popular general purpose tweezer used without magnification. It is made of carbon tool steel with flame hardened tips. The overall Length: is 4.5in. The tips measure.018in wide by.010in high.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TWEEZERS STRONG MEDIUM PNT CARBOFB 4.5IN 75880-762 00' Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty